Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrphostin AG 537, Thermo Scientific™

Catalog No. AAJ65263MA
Click to view available options
Quantity:
10 mg

Competitively inhibits epidermal growth factor receptor kinase autophosphorylation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 276881-63-1
Molecular Formula C23H20N4O6
MDL Number MFCD00236440
Synonym Bis-Tyrphostin

Specifications

Color Yellow
Quantity 10 mg
Sensitivity Light sensitive
Formula Weight 448.43
Physical Form Powder
Chemical Name or Material Tyrphostin AG 537
TSCA No
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.