Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TYSND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556435
Description
Immunogen sequence: ATQETCPYDI AVVSLEEDLD DVPIPVPAEH FHEGEAVSVV GFGVFGQSCG PSVTSGILSA VVQVNGTPVM LQTTCAV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Specifications
| TYSND1 | |
| Polyclonal | |
| Unconjugated | |
| TYSND1 | |
| 1300019N10Rik; NET41; peroxisomal cysteine endopeptidase; peroxisomal leader peptide-processing protease; Peroxisomal leader peptide-processing protease, 10 kDa form; Peroxisomal leader peptide-processing protease, 15 kDa form; Peroxisomal leader peptide-processing protease, 45 kDa form; Peroxisomal leader peptide-processing protease, 49 kDa form; trypsin domain containing 1; trypsin domain-containing protein 1; Tysnd1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219743, 365571 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2T9J0 | |
| TYSND1 | |
| Recombinant protein corresponding to Human TYSND1. Recombinant protein control fragment (Product #RP-95863). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction