Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Comfort Fit Design 127S Series Coveralls, NAFTA
GREENER_CHOICE

Catalog No. 17500238
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Meets NAFTA Standards.

  • Tyvek 400 coveralls provide an ideal balance of protection, durability and comfort of any limited use fabric technology.
  • Fabric offers an inherent barrier against particles (down to 1.0 micron in size).
  • Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
  • Comfort fit design based on extensive wear.
  • Enables a greater range of movement while stretching and bending, provides a more tailored fit.
  • Longer zipper for easier donning and doffing and an elastic waist to better position the garment.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Elastic opening for tighter fit at wrist, elastic opening for tighter fit at ankle
  • Model 127 USMCA sourced
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Latex Free Yes
Model TY127SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.