Learn More
DuPont™ Tyvek™ 400 Disposable Shirt
GREENER_CHOICE
Catalog No. p-7139001
Click to view available options
Size:
5X-Large
6X-Large
7X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
DuPont™ Tyvek™ 400 shirts provide the protection and durability you need so you can focus on the task at hand.
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
TRUSTED_SUSTAINABILITY
Specifications
Size 5X-Large
Color White
Neck Style Collar
Seam Type Serged
Closure Type Snaps
Cuff Style Open
Style Extends to hip
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.