Learn More
DuPont™ Tyvek™ 500 HV Coveralls with Hood
Catalog No. 17000542
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Tyvek™ 500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort. Thanks to their nonwoven structure, they are permeable to both air and water vapor, yet repel water-based liquids and aerosols.
  • Collared coverall available in fluorescent orange with silver grey retroreflective bands for day and night visibility, available in sizes SM to 3X.
  • Robust yet lightweight.
  • Mandarin collar, elasticated wrists and ankles as well as glued-in waist elastic.
  • Ample crotch area for freedom of movement.
  • Large, easy-to grasp zipper puller.
  • Tyvek™ 500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort.
    • Thanks to their nonwoven structure, they are permeable to both air and water vapor, yet repel water-based liquids and aerosols.
    • They offer an excellent barrier against fine particles and fibers, are ultra-low linting and antistatically treated on the inside.
    • Their bright fluorescent orange color background material combined with silver grey retroreflective material make them highly visible during the day as well as in the night (when exposed to a light source).
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3).
  • Antistatic treatment (EN 1149-5) - on inside.
  • Stitched external seams.
  • Very low inward leakage thanks to optimized design.
  • Tyvek™ auto-lock zipper and zipper flap for increased protection.
  • Tyvek™ 500 HV meets the single-use disposable coverall requirements of ANSI/ISEA 107-2020 Type R/Class 3 Section 11
TRUSTED_SUSTAINABILITY
Specifications
Product Type High Visibility Coveralls
Size 5X-Large
Material Polyethylene
Color Orange, Gray
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type Respirator Fit
Closure Type Zipper
Seam Type Serged
Certifications/Compliance ANSI/ISEA 107-2020 Type R/Class 3 Section 11
Certifications/Compliance (Europe) EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3), Antistatic treatment (EN 1149-5) on inside
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.