Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Extra Large Bag
Designed for the steam sterilization of large or odd shaped pharmaceutical equipment change parts. These bags are closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
$3458.00 - $7392.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | COC |
Closure Type | Drawstring |
Color | White |
Disposable | Single-use |
Catalog Number | Mfr. No. | Dimensions (L x W) | Capacity (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Capacity (English) | Price | Quantity & Availability | ||||
19-990-354
![]() |
Keystone Cleanroom Products
BGT2A1072D |
72 x 10 in. | 648 oz. |
Case of 250 for $3,861.00
|
|
||||
19-990-375
![]() |
Keystone Cleanroom Products
BGT2A1448D |
48 x 14 in. | 624 oz. |
Case of 250 for $4,243.00
|
|
||||
19-990-376
![]() |
Keystone Cleanroom Products
BGT2A1543D |
43 x 15 in. | 602 oz. |
Case of 250 for $3,458.00
|
|
||||
19-990-384
![]() |
Keystone Cleanroom Products
BGT2A1644D |
44 x 16 in. | 660 oz. |
Case of 250 for $3,668.00
|
|
||||
19-990-385
![]() |
Keystone Cleanroom Products
BGT2A1648D |
48 x 16 in. | 720 oz. |
Case of 250 for $3,737.00
|
|
||||
19-990-391
![]() |
Keystone Cleanroom Products
BGT2A1836D |
36 x 18 in. | 612 oz. |
Case of 250 for $3,564.00
|
|
||||
19-990-392
![]() |
Keystone Cleanroom Products
BGT2A1840D |
40 x 18 in. | 680 oz. |
Case of 250 for $3,682.00
|
|
||||
19-990-393
![]() |
Keystone Cleanroom Products
BGT2A1846D |
46 x 18 in. | 782 oz. |
Case of 250 for $3,781.00
|
|
||||
19-990-394
![]() |
Keystone Cleanroom Products
BGT2A1848D |
48 x 18 in. | 816 oz. |
Case of 250 for $3,861.00
|
|
||||
19-990-395
![]() |
Keystone Cleanroom Products
BGT2A1854D |
54 x 18 in. | 918 oz. |
Case of 250 for $4,097.00
|
|
||||
19-990-400
![]() |
Keystone Cleanroom Products
BGT2A2032D |
32 x 20 in. | 608 oz. |
Case of 250 for $3,616.00
|
|
||||
19-990-401
![]() |
Keystone Cleanroom Products
BGT2A2036D |
36 x 20 in. | 684 oz. |
Case of 250 for $3,733.00
|
|
||||
19-990-402
![]() |
Keystone Cleanroom Products
BGT2A2040D |
40 x 20 in. | 760 oz. |
Case of 250 for $3,616.00
|
|
||||
19-990-403
![]() |
Keystone Cleanroom Products
BGT2A2044D |
44 x 20 in. | 836 oz. |
Case of 250 for $4,070.00
|
|
||||
19-990-404
![]() |
Keystone Cleanroom Products
BGT2A2046D |
46 x 20 in. | 874 oz. |
Case of 250 for $4,147.00
|
|
||||
19-990-405
![]() |
Keystone Cleanroom Products
BGT2A2048D |
48 x 20 in. | 912 oz. |
Case of 250 for $4,266.00
|
|
||||
19-990-407
![]() |
Keystone Cleanroom Products
BGT2A2436D |
36 x 24 in. | 828 oz. |
Case of 250 for $4,411.00
|
|
||||
19-990-408
![]() |
Keystone Cleanroom Products
BGT2A2440D |
40 x 24 in. | 920 oz. |
Case of 250 for $4,512.00
|
|
||||
19-990-410
![]() |
Keystone Cleanroom Products
BGT2A2634D |
34 x 26 in. | 850 oz. |
Case of 250 for $4,800.00
|
|
||||
19-990-412
![]() |
Keystone Cleanroom Products
BGT2A2828D |
28 x 28 in. | 756 oz. |
Case of 250 for $4,199.00
|
|
||||
19-990-413
![]() |
Keystone Cleanroom Products
BGT2A2832D |
32 x 28 in. | 864 oz. |
Case of 250 for $7,392.00
|
|
||||
19-990-414
![]() |
Keystone Cleanroom Products
BGT2A3028D |
30 x 28 in. | 812 oz. |
Case of 250 for $5,096.00
|
|
||||
19-990-415
![]() |
Keystone Cleanroom Products
BGT2A3030D |
30 x 30 in. | 870 oz. |
Case of 250 for $5,353.00
|
|
||||
19-990-416
![]() |
Keystone Cleanroom Products
BGT2A3232D |
32 x 32 in. | 992 oz. |
Case of 250 for $5,436.00
|
|
||||
19-990-419
![]() |
Keystone Cleanroom Products
BGT2A3628D |
36 x 28 in. | 980 oz. |
Case of 250 for $5,740.00
|
|
||||
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams eliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Specifications
Autoclavable | |
Drawstring | |
Single-use | |
Pharmaceutical Equipment | |
Non-sterile | |
Steam | |
<125°C |
COC | |
White | |
Steam Sterilization | |
Tyvek | |
Yes | |
<250°F | |
Autoclave Bag |