Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ Isoclean™ 255 Series Clean and Sterile Coveralls



Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Description
- Tyvek flashspun polyolefin protective material
- Patented flash-spinning process
- Creates barrier to dry particles, microorganisms, non-hazardous liquids
- Lightweight, durable
- Collared

Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | Medium |
Gender | Unisex |
Material | Tyvek™ |
Disposable | Single-use |
Hood Type | Standard |
Seam Type | Serged |
Sterility | Sterile |
Certifications/Compliance (Europe) | CE |
Model | IC255BWH |
Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction