Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TYW1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553065 Supplier Invitrogen™ Supplier No. PA553065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEQQEEKSGL FRNMGRNEDG ERRAMITPAL REALTKQVDA PRERSLLQTH ILW Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 49%.

Wybutosine is a hypermodified guanosine found in phenylalanine tRNA. Wybutosine functions to stabilize codon-anticodon interactions during ribosome decoding and therefore supports the maintenance of the reading frame. In yeast, the homolog of this gene is essential for the synthesis of wybutosine. The human genome contains two closely related genes that putatively function in wybutosine synthesis. The open reading frame of this locus is disrupted in some individuals. Thus, this locus appears to be an evolving pseudogene, but may still be functional in some members of the population. [provided by RefSeq, Apr 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYW1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TYW1B
Gene Accession No. Q6NUM6
Gene Alias LINC00069; long intergenic non-protein coding RNA 69; MGC87315; NCRNA00069; radical S-adenosyl methionine and flavodoxin domain-containing protein 2; radical S-adenosyl methionine and flavodoxin domains 1; RSAFD2; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase TYW1B; tRNA wybutosine-synthesizing protein 1 homolog B; tRNA-yW synthesizing protein 1 homolog B; tRNA-yW synthesizing protein 1 homolog B (non-protein coding); TYW1B
Gene Symbols TYW1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TYW1. Recombinant protein control fragment (Product #RP-102540).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.