Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TYW1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553316 Supplier Invitrogen™ Supplier No. PA553316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRAMITPALR EALTKQGYQL IGSHSGVKLC RWTKSMLRGR GGCYKHTFYG IESHRCMETT PSLACANKCV FCWRHHTNPV GTEWRWKMDQ PEMILKEAIE NHQNMIKQFK GVPGVKA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

TYW1, also known as TYW1A, RSAFD1 or YPL207W, is a 732 amino acid protein containing one flavodoxin-like domain that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW1 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation. TYW1 binds to one 4Fe-4S cluster and is located on human chromosome 7. Defects in some of the genes on chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome, suggesting that TYW1 may play a role in these conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYW1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TYW1
Gene Accession No. Q9NV66
Gene Alias Radical S-adenosyl methionine and flavodoxin domain-containing protein 1; radical S-adenosyl methionine and flavodoxin domains 1; RGD1310420; RSAFD1; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase TYW1; tRNA wybutosine-synthesizing protein 1 homolog; tRNA-yW synthesizing protein 1 homolog; tRNA-yW synthesizing protein 1 homolog (S. cerevisiae); tRNA-yW synthesizing protein 1 homolog A; tRNA-yW-synthesizing protein; TYW1; TYW1A; YPL207W
Gene Symbols TYW1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TYW1. Recombinant protein control fragment (Product #RP-90614).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.