Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TYW3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556221 Supplier Invitrogen™ Supplier No. PA556221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFRKWKAQCL SKADLSRKGS VDEDVVELVQ FLNMRDQFFT TSSCAGRILL LDRGINGFEV QKQNCCWLLV THKLCVKDDV IV Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 77%.

TYW3 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW3 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TYW3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TYW3
Gene Accession No. Q6IPR3
Gene Alias 5230400J09Rik; C1orf171; hypothetical protein BC009514; LOC519731 protein; RGD1559623; tRNA wybutosine-synthesizing protein 3 homolog; tRNA(Phe) 7-((3-amino-3-carboxypropyl)-4-demethylwyosine(37)-N(4))-methyltransferase; tRNA-yW synthesizing protein 3 homolog; tRNA-yW synthesizing protein 3 homolog (S. cerevisiae); tRNA-yW-synthesizing protein 3; Tyw3
Gene Symbols TYW3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TYW3. Recombinant protein control fragment (Product #RP-94393).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.