Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TZFP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | TZFP |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TZFP Polyclonal antibody specifically detects TZFP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TZFP | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Fanconi anemia zinc finger protein, FAXF, FAZFFANCC-interacting protein, ROG, Testis zinc finger protein, TZFPZinc finger protein 538, zinc finger and BTB domain containing 32, zinc finger and BTB domain-containing protein 32, ZNF538FAXFrepressor of GATA | |
This antibody was developed against a recombinant protein corresponding to amino acids: CPSLASMQAHMRGHSPSQLPPGWTIRSTFLYSSSRPSRPSTSPCCPSSSTT | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol | |
27033 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title