Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TZFP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518204 Supplier Invitrogen™ Supplier No. PA518204
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

DNA-binding protein that binds to the to a 5'-TGTACAGTGT-3' core sequence. May function as a transcriptional transactivator and transcriptional repressor. Probably exerts its repressor effect by preventing GATA3 from binding to DNA. May play a role in regulating the differentiation and activation of helper T-cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TZFP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ZBTB32
Gene Accession No. Q9Y2Y4
Gene Alias 4930524C15Rik; FANCC-interacting protein; Fanconi anemia zinc finger protein; FAXF; FAZF; GATA repressor; OTTHUMP00000045790; PLZP; repressor of GATA; Rog; Testis zinc finger protein; TZFP; ZBTB32; zinc finger and BTB domain containing 32; zinc finger and BTB domain-containing protein 32; Zinc finger protein 538; ZNF538
Gene Symbols ZBTB32
Host Species Goat
Immunogen Synthetic peptide sequence (RPSTSPCCPSSSTT) corresponding to the C-terminus amino acids of ZBTB32 (aa 474-487).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27033
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.