Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TZFP (ZBTB32) Monoclonal Antibody (OTI2H8), TrueMAB™, OriGene
SDP

Catalog No. 502543768
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3768 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3768 Supplier OriGene Supplier No. CF812203
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TZFP (ZBTB32) Monoclonal antibody specifically detects TZFP (ZBTB32) in Human samples. It is validated for Western Blot

Specifications

Antigen TZFP (ZBTB32)
Applications Western Blot
Classification Monoclonal
Clone OTI2H8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene COX17
Gene Accession No. Q14061
Gene Alias 4930524C15Rik; FANCC-interacting protein; Fanconi anemia zinc finger protein; FAXF; FAZF; GATA repressor; OTTHUMP00000045790; PLZP; repressor of GATA; Rog; Testis zinc finger protein; TZFP; ZBTB32; zinc finger and BTB domain containing 32; zinc finger and BTB domain-containing protein 32; Zinc finger protein 538; ZNF538
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-63 of human COX17 (NP_005685) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10063
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.