Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ U 46619
GREENER_CHOICESDP

Catalog No. 19321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-321 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-321 Supplier Tocris Bioscience™ Supplier No. 1932/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, stable thromboxane A2 (TP) receptor agonist

PGH2 (TXA2) analog that is a potent and stable thromboxane A2 (TP) receptor agonist (EC50 = 0.035 μM). Potently stimulates TP receptor-mediated, but not other prostaglandin receptor-mediated responses in various in vitro preparations. Activates ERK-1 and ERK-2 in HEK 293 cells expressing TPα and TPβ receptors.

Chemical Identifiers

CAS 56985-40-1
Molecular Formula C21H34O4

Specifications

Quantity 1 mg
Formula Weight 350.5
Purity >98%
Molecular Weight (g/mol) 350.499
Chemical Name or Material U 46619
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.