Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U-47700-D3 solution, 100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant

Catalog No. U0041ML Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mL

This stable-labeled Snap-N-Shoot internal standard is offered as DEA-exempt solution and suitable for quantitation of U-47700 levels in urine or blood by LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications.

This stable-labeled Snap-N-Shoot™ internal standard is offered as DEA-exempt solution and suitable for quantitation of U-47700 levels in urine or blood by LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. U-47700 is a highly potent opioid agonist, not structurally related to fentanyl, that has been confirmed in opioid overdoses and deaths. Novel synthetic opioids (NSOs) created to circumvent drug control laws such as U-47700 have no recognized therapeutic use, are clandestinely manufactured and sold on conventional or dark web.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material U-47700-D3
Packaging Ampule of 1 mL
Molecular Formula C16H19D3Cl2N2O
Concentration 100 μg/mL in Methanol
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Molecular Weight (g/mol) 332.28
Industry Type Clinical Testing
Quantity 1 mL
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.