Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U-54494A hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
κ-opioid agonist and anticonvulsant. Perhaps acts as an NMDA antagonist.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 391.77 |
Percent Purity | >98% |
Chemical Name or Material | U-54494A hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction