Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ U 73122
GREENER_CHOICESDP

Catalog No. 126850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-685-0 50 mg
12-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-685-0 Supplier R&D Systems™ Supplier No. 1268/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Phospholipase C inhibitor

Phospholipase C inhibitor. Inhibits agonist-induced platelet aggregation with IC50 values of 1-5 μM. Potently inhibits human polymorphonuclear neutrophil adhesion on biological surfaces (IC50 < 50 nM) and exhibits antinociceptive activity in vivo. Also activates TRPM4 and inhibits TRPM3 channels.

Negative Control also available.

Chemical Identifiers

CAS 112648-68-7
Molecular Formula C29H40N2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 464.65
Purity >98%
Molecular Weight (g/mol) 464.65
Chemical Name or Material U 73122
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.