Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ U 73343
GSA_VAGREENER_CHOICE

Catalog No. 413310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-331-0 10 mg
41-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-331-0 Supplier Tocris Bioscience™ Supplier No. 4133/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Analog of U 73122 (Cat. No. 1268)

Analog of U 73122 ; can be used as a negative control. Inhibits Panx1 currents in HEK cells. Also inhibits vasopressin- and GTPγS-induced Ca2+ influx in hepatocytes.

Active Analog also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 142878-12-4
Molecular Formula C29H42N2O3

Specifications

Quantity 10 mg
Formula Weight 466.66
Purity >98%
Molecular Weight (g/mol) 466.67
Chemical Name or Material U 73343
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.