Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U1A Rabbit anti-Human, Mouse, Rat, Clone: 8I3P5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | U1A |
---|---|
Clone | 8I3P5 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
U1A Monoclonal antibody specifically detects U1A in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
U1A | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6626 | |
IgG | |
Affinity purified |
8I3P5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Mud1, small nuclear ribonucleoprotein polypeptide A, U1 snRNP A, U1 snRNP-specific protein A, U1-AU1 small nuclear ribonucleoprotein A, U1AU1 small nuclear RNP-specific A | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human U1A (P09012). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title