Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U2AF1L4 Monoclonal Antibody (OTI7B12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810433

Description
RNA-binding protein that function as a pre-mRNA splicing factor. Plays a critical role in both constitutive and enhancer-dependent splicing by mediating protein-protein interactions and protein-RNA interactions required for accurate 3′-splice site selection. Acts by enhancing the binding of U2AF2 to weak pyrimidine tracts. Also participates in the regulation of alternative pre-mRNA splicing. Activates exon 5 skipping of PTPRC during T cell activation; an event reversed by GFI1. Binds to RNA at the AG dinucleotide at the 3′-splice site.Specifications
U2AF1L4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WU68 | |
U2AF1L4 | |
Full length human recombit protein of human U2AF1L4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
OTI7B12 | |
Unconjugated | |
U2AF1L4 | |
U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26 | |
Mouse | |
Affinity Chromatography | |
RUO | |
199746 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction