Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ U2AF1L4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518826
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518826 Supplier Invitrogen™ Supplier No. PA518826
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

RNA-binding protein that function as a pre-mRNA splicing factor. Plays a critical role in both constitutive and enhancer-dependent splicing by mediating protein-protein interactions and protein-RNA interactions required for accurate 3'-splice site selection. Acts by enhancing the binding of U2AF2 to weak pyrimidine tracts. Also participates in the regulation of alternative pre-mRNA splicing. Activates exon 5 skipping of PTPRC during T-cell activation; an event reversed by GFI1. Binds to RNA at the AG dinucleotide at the 3'-splice site. Shows a preference for AGC or AGA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen U2AF1L4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene U2AF1L4
Gene Accession No. Q8WU68
Gene Alias FLJ35525; MGC33901; Splicing factor U2AF 26 kDa subunit; U2 auxiliary factor 26; U2 small nuclear RNA auxiliary factor 1 like 4; U2 small nuclear RNA auxiliary factor 1-like 3; U2 small nuclear RNA auxiliary factor 1-like 4; U2 small nuclear RNA auxiliary factor 1-like protein 3; U2 small nuclear RNA auxiliary factor 1-like protein 4; U2(RNU2) small nuclear RNA auxiliary factor 1-like 3; U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3; U2AF1L3; U2AF1L3V1; U2AF1L4; U2AF1-like 4; U2AF1-like protein 3; U2AF1RS3; U2AF1-RS3; U2af26
Gene Symbols U2AF1L4
Host Species Goat
Immunogen Synthetic peptide sequence (EDGERAVAELSNR) corresponding to the internal amino acids of U2AF1L4 (aa 121-134).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199746
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.