Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U2AF1L4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | U2AF1L4 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
U2AF1L4 Polyclonal specifically detects U2AF1L4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
U2AF1L4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
199746 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
FLJ35525, MGC33901, splicing factor U2AF 26 kDa subunit, U2 auxiliary factor 26, U2 small nuclear RNA auxiliary factor 1-like 3, U2 small nuclear RNA auxiliary factor 1-like 4, U2 small nuclear RNA auxiliary factor 1-like protein 3, U2 small nuclear RNA auxiliary factor 1-like protein 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 3, U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3, U2AF1L3, U2AF1L3V1, U2AF1-like protein 3, U2AF1RS3, U2AF1-RS3, U2af26 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-202 of human U2AF1L4 (NP_659424.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title