Learn More
U2AF35 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302079AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
U2AF35 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q01081, Q9D883 | |
U2af1 | |
Between 140 and 190 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
U2AF1 | |
Splicing factor U2AF 35 kDa subunit; U2 auxiliary factor 35 kDa subunit; U2 snRNP auxiliary factor small subunit; U2AF1; U2AF35; U2AFBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108121, 7307 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.