Learn More
U2SURP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | U2SURP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8200
![]() |
Bethyl Laboratories, Inc
A304616AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
The specific function of SR140 is not yet known.Specifications
U2SURP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
140 kDa Ser/Arg-rich domain protein, fSAPa, functional spliceosome-associated protein a, Ser/Arg-rich domain protein, 140 kDa, SR140, U2 associated SR140 protein, U2 snRNP-associated SURP domain containing, U2-associated protein SR140, U2-associated SR140 protein | |
Between 1 and 50 | |
Primary | |
4° C | |
U2SURP |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O15042, Q6NV83 | |
23350, 67958 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.