Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U2SURP Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501568198
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8198 Supplier Bethyl Laboratories, Inc Supplier No. A304616A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan

The specific function of SR140 is not yet known.

Specifications

Antigen U2SURP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene U2SURP
Gene Accession No. O15042, Q6NV83
Gene Alias 140 kDa Ser/Arg-rich domain protein, fSAPa, functional spliceosome-associated protein a, Ser/Arg-rich domain protein, 140 kDa, SR140, U2 associated SR140 protein, U2 snRNP-associated SURP domain containing, U2-associated protein SR140, U2-associated SR140 protein
Gene Symbols U2SURP
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23350, 67958
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.