Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UACA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559560 Supplier Invitrogen™ Supplier No. PA559560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELNKQLKDLS QKYTEVKNVK EKLVEENAKQ TSEILAVQNL LQKQHVPLEQ VEALKKSLNG TIENLKEELK SMQRCYEKEQ QTVTKLHQLL ENQKNSSVPL A Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.

Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UACA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UACA
Gene Accession No. Q9BZF9
Gene Alias 2700059D02Rik; beta-actin binding protein betaCAP73; beta-actin-binding protein; BetaCAP73; Kiaa1561; nuclear membrane binding protein; nuclear membrane binding protein NUCLING; nuclear membrane-binding protein; Nucling; RGD1560011; Uaca; uveal autoantigen; uveal autoantigen with coiled coil domains and ankyrin repeats; uveal autoantigen with coiled-coil domains and ankyrin repeats; Uveal autoantigen with coiled-coil domains and ankyrin repeats protein
Gene Symbols UACA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UACA. Recombinant protein control fragment (Product #RP-98066).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.