Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UACA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987194
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7194 Supplier Bioss Supplier No. BS6308R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Regulates APAF1 expression and plays an important role in the regulation of stress-induced apoptosis. Promotes apoptosis by regulating three pathways, apoptosome up-regulation, LGALS3/galectin-3 down-regulation and NF-kappa-B inactivation. Regulates the redistribution of APAF1 into the nucleus after proapoptotic stress. Down-regulates the expression of LGALS3 by inhibiting NFKB1.

Specifications

Antigen UACA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UACA
Gene Accession No. Q9BZF9
Gene Alias 2700059D02Rik; Kiaa1561; nuclear membrane binding protein; nuclear membrane binding protein NUCLING; Nuclear membrane-binding protein; Nucling; RGD1560011; UACA; uveal autoantigen with coiled coil domains and ankyrin repeats; Uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Symbols UACA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UACA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55075
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.