Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UAP1 Antibody (SR2041), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | UAP1 |
---|---|
Clone | SR2041 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
UAP1 Monoclonal antibody specifically detects UAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
UAP1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AGX, AGX1, AGX2, Antigen X, EC 2.7.7, EC 2.7.7.23, EC 2.7.7.27, SPAG2, sperm associated antigen 2, Sperm-associated antigen 2, UDP-N-acetylhexosamine pyrophosphorylase, UDP-N-acteylglucosamine pyrophosphorylase 1 | |
A synthesized peptide derived from human UAP1 (Uniprot #: Q16222) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2041 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6675 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title