Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UAP1 Monoclonal Antibody (4H1)
GREENER_CHOICE

Catalog No. PIMA534892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534892 Supplier Invitrogen™ Supplier No. MA534892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: Jurkat, A549, Hela, rat epididymis tissue, human lung cancer tissue, human colon cancer tissue, mouse testis tissue.

Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene UAP1
Gene Accession No. Q16222
Gene Alias AA420407; AA437972; AgX; AGX1; AGX-1; AGX2; AGX-2; antigen X; ESTM38; fb63d12; RGD1561967; similar to UDP-N-acetylhexosamine pyrophosphorylase (Antigen X) (AGX) (Sperm-associated antigen 2); SPAG2; sperm associated antigen 2; sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UAP1; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylglucosamine pyrophosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1; UDP-N-acteylglucosamine pyrophosphorylase 1; wu:fb63d12; zgc:55280
Gene Symbols UAP1
Host Species Mouse
Immunogen Synthetic peptide within Human UAP1 aa 33-82.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6675
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.