Learn More
UAP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305642AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.Specifications
UAP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q16222 | |
UAP1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
UAP1 | |
AGX, AGX1, AGX2, SPAG2, UDP-N-acetylhexosamine pyrophosphorylase, antigen X, sperm associated antigen 2, sperm-associated antigen 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6675 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.