Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UAP1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560688 Supplier Invitrogen™ Supplier No. PA560688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPNWTLDPEP RCRLWSEPRL PAGPGVLAAG SPRLPCRYVM TSEFTLGPTA EFFREHNFFH LDPANVVMFE QRLLPAVTFD GKVILER Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 47%.

UAP1 is a 56KDa isoform which catalyzes the final step of UDP-N-acetylglucosamine (UDP-GlcNAc) biosynthesis from fructose-6-phosphate. UDP-GlcNAc, the activated form of GlcNAc moiety is used as a key precursor in N-linked glycosylation and the glycosylphosphatidylinositol (GPI)-anchor of cellular proteins. Crystallization studies revealed UAP1 is composed of a large central core flanked by two smaller extra domains. The central main domain displays a alpha/beta structure resembling the Rossmann fold. Northern Blot analysis detected abundant expression of UAP1 in testis and sperm with a low expression in placenta, muscle and liver. It is implicated in antibody-mediated infertility in males.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UAP1L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UAP1L1
Gene Accession No. Q3KQV9
Gene Alias 5730445F03Rik; UAP1; uap1l1; UDP-N-acetylglucosamine pyrophosphorylase 1 like 1; UDP-N-acetylglucosamine pyrophosphorylase 1, like 1; UDP-N-acetylhexosamine pyrophosphorylase-like protein 1; UDP-N-acteylglucosamine pyrophosphorylase 1, like 1; UDP-N-acteylglucosamine pyrophosphorylase 1-like 1; zgc:56509
Gene Symbols UAP1L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UAP1L1. Recombinant protein control fragment (Product #RP-93117).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91373
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.