Learn More
Invitrogen™ UAP56 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Xenopus laevis (96%), Dog (100%), Pig (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | UAP56 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DDX39B |
| Gene Accession No. | Q13838, Q63413, Q9Z1N5 |
| Gene Alias | 0610030D10Rik; 56 kDa U2AF65-associated protein; AI428441; ATP-dependent RNA helicase p47; Bat1; Bat-1; Bat1a; D17H6S81E; D17H6S81E-1; D20H6S81e; D6S81E; D6S81Eh; Ddx39b; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B; DEAD box protein UAP56; DEAD-box helicase 39B; DExD-box helicase 39B; HLA-B associated transcript 1; HLA-B associated transcript 1A; HLA-B-associated transcript 1 protein; HLA-B-associated transcript 1A; Nuclear RNA helicase; nuclear RNA helicase (DEAD family); nuclear RNA helicase Bat1; p47; spliceosome RNA helicase BAT1; spliceosome RNA helicase DDX39B; Uap56 |
| Gene Symbols | DDX39B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.