Learn More
Thermo Scientific™ UAS-bla HEK 293T Master Cell Line
Description
The UAS-bla HEK 293T Master Cell Line consists of HEK 293T cells stably transfected with a UAS-bla fusion construct. This cell line can be used to build cell-based assays by transfecting with GAL4 fusion constructs. Activation of the GAL4 fusion results in binding to the UAS-bla and subsequent expression of beta lactamase. Expression of beta lactamase is monitored using a FRET-based assay.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | Shipping: Dry Ice Storage: Liquid Nitrogen |
| Detection Method | Fluorescence |
| Cell Line | 293T (HEK) |
| Culture Type | Adherent Cell Culture |
| No. of Cells | 2 x 106 Cells |
| Product Line | GeneBLAzer |
| Product Type | HEK 293T Cells |
| Quantity | 2 x 106 cells |
| Reporter Gene | BLA (Beta-Lactamase) |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.