Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ub-K63 Antibody (HWA4C4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200199AF405
Description
Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.Specifications
Ub-K63 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
The immunogen for this antibody is Ub-K63. | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot | |
HWA4C4 | |
Western Blot | |
Mouse | |
Protein A or G purified | |
RUO | |
HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction