Learn More
UBA1 Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human UBA1.
Supplier: Abnova Corporation PAB29508
Description
The protein encoded by this gene catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternatively spliced transcript variants that encode the same protein have been described. [provided by RefSeq]
Sequence: LAAPRHQYYNQEWTLWDRFEVQGLQPNGEEMTLKQFLDYFKTEHKLEITMLSQGVSMLYSFFMPAAKLKERLDQPMTEIVSRVSKRKLGRHVRALVLELCCNDESGEDVEVPSpecifications
UBA1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human UBA1. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
UBA1 | |
Recombinant protein corresponding to human UBA1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
A1S9/A1S9T/A1ST/AMCX1/GXP1/MGC4781/SMAX2/UBA1A/UBE1/UBE1X | |
Rabbit | |
Antigen affinity purification | |
RUO | |
7317 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.