Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBA2 Monoclonal Antibody (OTI4A3)
GREENER_CHOICE

Catalog No. PIMA525437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525437 Supplier Invitrogen™ Supplier No. MA525437
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBA2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBA2
Gene Accession No. Q9UBT2
Gene Alias AA986091; Anthracycline-associated resistance ARX; Arx; EGK_10449; FLJ13058; HRIHFB2115; SAE2; SUMO1 activating enzyme subunit 2; Sumo-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; UBA1; UBA2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin like modifier activating enzyme 2; ubiquitin-like 1 (sentrin) activating enzyme E1B; ubiquitin-like 1 (sentrin) activating enzyme subunit 2; ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; ubiquitin-like modifier-activating enzyme 2; Ubl1a2; Uble1b
Gene Symbols UBA2
Host Species Mouse
Immunogen Full length human recombinant protein of UBA2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10054, 308508, 476490
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.