Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575187 Supplier Invitrogen™ Supplier No. PA575187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene UBA2
Gene Accession No. Q9UBT2, Q9Z1F9
Gene Alias AA986091; Anthracycline-associated resistance ARX; Arx; EGK_10449; FLJ13058; HRIHFB2115; SAE2; SUMO1 activating enzyme subunit 2; Sumo-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; UBA1; UBA2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin like modifier activating enzyme 2; ubiquitin-like 1 (sentrin) activating enzyme E1B; ubiquitin-like 1 (sentrin) activating enzyme subunit 2; ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; ubiquitin-like modifier-activating enzyme 2; Ubl1a2; Uble1b
Gene Symbols UBA2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 598-640 of Human SAE2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10054, 308508, 50995
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.