Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA511194
Description
UBA2 Polyclonal Antibody for Western Blot, IHC (P)
UBA2 is involved in covalent attachment of one protein to another is one of the more prominent posttranslational modifications in respect to size and ubiquity. Ubiquitin is the most familiar of the protein modifiers and its activation and transfer to target proteins has been studied extensively. Recently, a new group of ubiquitin-like proteins has been receiving a lot of attention. SUMO, or Sentrin, is one of the most intriguing. SUMO has been implicated in the stabilization of target proteins and/or their localization to sub-cellular complexes.The conjugation of SUMO-1, SUMO-2 and SUMO-3 onto target proteins requires the concerted action of the specific E1-activating enzyme SAE1/SAE2, the E2-conjugating enzyme Ubc9, and an E3-like SUMO ligase.
Specifications
UBA2 | |
Polyclonal | |
Unconjugated | |
UBA2 | |
AA986091; Anthracycline-associated resistance ARX; Arx; EGK_10449; FLJ13058; HRIHFB2115; SAE2; SUMO1 activating enzyme subunit 2; Sumo-1 activating enzyme subunit 2; SUMO-activating enzyme subunit 2; UBA1; UBA2; UBA2, ubiquitin-activating enzyme E1 homolog; ubiquitin like modifier activating enzyme 2; ubiquitin-like 1 (sentrin) activating enzyme E1B; ubiquitin-like 1 (sentrin) activating enzyme subunit 2; ubiquitin-like 1-activating enzyme E1B; ubiquitin-like modifier activating enzyme 2; ubiquitin-like modifier-activating enzyme 2; Ubl1a2; Uble1b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UBT2 | |
UBA2 | |
KLH conjugated synthetic peptide between 601-631 amino acids from the C-terminal region of human SAE2 (UBA2). | |
400 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction