Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBA3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $648.50
Specifications
Antigen | UBA3 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
UBA3 Polyclonal specifically detects UBA3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
UBA3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
DKFZp566J164, EC 6.3.2, EC 6.3.2.-, hUba3, MGC22384, NEDD8-activating enzyme E1 catalytic subunit, NEDD8-activating enzyme E1C, Nedd8-activating enzyme hUba3, UBA3, ubiquitin-activating enzyme E1 homolog, UBE1C, Ubiquitin-activating enzyme 3, Ubiquitin-activating enzyme E1C, ubiquitin-activating enzyme E1C (homologous to yeast UBA3), ubiquitin-like modifier activating enzyme 3, Ubiquitin-like modifier-activating enzyme 3, yeast) | |
UBA3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cell Cycle and Replication, Neuroscience | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9039 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KIATSAYIPLNNYLVFNDVDGLYTYTFEAERKENCPACSQLPQNIQFSPSAKLQEVLDYLTNSASLQMKSPAITATLEGKNRTLYLQSVTSIEERTR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title