Learn More
UBA3 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1B10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811880

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E1 ubiquitin-activating enzyme family. The encoded enzyme associates with AppBp1, an amyloid beta precursor protein binding protein, to form a heterodimer, and then the enzyme complex activates NEDD8, a ubiquitin-like protein, which regulates cell division, signaling and embryogenesis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
UBA3 | |
Monoclonal | |
Unconjugated | |
UBA3 | |
NAE2, UBE1C, hUBA3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
117553, 22200, 476560, 9039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8C878, Q8TBC4, Q99MI7 | |
Uba3 | |
Human recombit protein fragment corresponding to amino acids 1-463 of human UBA3 produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.