Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583250 Supplier Invitrogen™ Supplier No. PA583250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPPQVNFPMC TIASMPRLPE HCIEYVRMLQ WPKEQPFGEG VPLDGDDPEH IQWIFQKSLE RASQYNIRGV TYRLTQGVVK RIIPAVASTN AVIAAVCA Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E1 ubiquitin-activating enzyme family. The encoded enzyme associates with AppBp1, an amyloid beta precursor protein binding protein, to form a heterodimer, and then the enzyme complex activates NEDD8, a ubiquitin-like protein, which regulates cell division, signaling and embryogenesis. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Uba3
Gene Accession No. Q8TBC4
Gene Alias A830034N06Rik; AI256736; AI848246; AW546539; DKFZp566J164; hUBA3; MGC22384; NAE2; NEDD8-activating enzyme E1 catalytic subuni-like protein; NEDD8-activating enzyme E1 catalytic subunit; NEDD8-activating enzyme E1 subunit 2; NEDD8-activating enzyme E1C; Nedd8-activating enzyme hUba3; Uba3; UBA3, ubiquitin-activating enzyme E1 homolog; UBE1C; ubiquitin like modifier activating enzyme 3; ubiquitin-activating enzyme 3; Ubiquitin-activating enzyme E1C; ubiquitin-activating enzyme E1C (homologous to yeast UBA3); ubiquitin-activating enzyme E1C (UBA3 homolog, yeast); ubiquitin-like modifier activating enzyme 3; ubiquitin-like modifier-activating enzyme 3; Unknown (protein for MGC:140052); wu:fb75e04; wu:fc37b11; zgc:55528
Gene Symbols Uba3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBA3. Recombinant protein control fragment (Product #RP-104232).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.