Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553565 Supplier Invitrogen™ Supplier No. PA553565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIGKLLLFDY DKVELANMNR LFFQPHQAGL SKVQAAEHTL RNINPDVLFE VHNYNITTVE NFQHFMDRIS NGGLEEGKPV DLVLSCVDNF EARMTINTAC NELGQTWMES GVSENAVSGH IQLIIPGESA CFACAPPLVV AANIDEKTL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A pseudogene located on chromosome 1 has also been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBA5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBA5
Gene Accession No. Q9GZZ9
Gene Alias 5730525G14Rik; AW240750; cb972; FLJ23251; FLJ23251UBA5; THIFP1; UBA5; UBA5, ubiquitin-activating enzyme E1 homolog; ube1dc1; ubiquitin like modifier activating enzyme 5; ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; ubiquitin-like modifier-activating enzyme 5; ubiquitin-like modifier-activating enzyme 5-like protein; UFM1-activating enzyme
Gene Symbols UBA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBA5. Recombinant protein control fragment (Product #RP-90003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.