Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590523 Supplier Invitrogen™ Supplier No. PA590523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAESVERLQQ RVQELERELA QERSLQVPRS GDGGGGRVRI EKMSSEVVDS NPYSRLMALK RMGIVSDYEK IRTFAVAIVG VGGVGSVTAE MLTRCGIGKL LLFDYDKVEL ANMNRLFFQP HQAGLSKVQA AEHTLRNINP DVLFEVHNYN ITTVENFQHF MDRISNGGLE EGKPVDLVLS CVDNFEARMT INTACNELGQ TWMESGVSEN AVSGHIQLII PGESACFACA PPLVVAANID EKTLKREGVC AASLPTTMGV VAGILVQNVL KFLLNFGTVS FYLGYNAMQD FFPTMSMKPN PQCDDRNCRK QQEEYKKKVA ALPKQEVIQE EEEIIHEDNE WGIELVSEVS EEELKNFSGP VPDLPEGITV AYTIPKKQED SVTELTVEDS GESLEDLMAK MKNM; Positive Samples: HT-29, A-549, Jurkat, HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A pseudogene located on chromosome 1 has also been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBA5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBA5
Gene Accession No. Q5M7A4, Q8VE47, Q9GZZ9
Gene Alias 5730525G14Rik; AW240750; cb972; FLJ23251; FLJ23251UBA5; THIFP1; UBA5; UBA5, ubiquitin-activating enzyme E1 homolog; ube1dc1; ubiquitin like modifier activating enzyme 5; ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; ubiquitin-like modifier-activating enzyme 5; ubiquitin-like modifier-activating enzyme 5-like protein; UFM1-activating enzyme
Gene Symbols UBA5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-404 of human UBA5 (NP_0790941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300968, 66663, 79876
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.