Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBA52 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300263AF750
Description
UBA52 Polyclonal antibody specifically detects UBA52 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
UBA52 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
The immunogen this antibody was made to, maps to a region between residues 1 and 50 of human Ubiquitin using the numbering given in entry P62988.(GeneID 7311). | |
0.1 mL | |
Apoptosis, Cell Biology | |
7311 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction