Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBA52 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | UBA52 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UBA52 Polyclonal specifically detects UBA52 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
UBA52 | |
Polyclonal | |
Rabbit | |
Human | |
CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
UBA52 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
7311 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RGGIIEPSLRQLAQKYNCDKMICRKCYARLHPRAVNCRKKKCGHTNNLRPKKKVK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title