Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBA52 Antibody (OTI4E1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274729H
Description
UBA52 Monoclonal specifically detects UBA52 in Human, Mouse samples. It is validated for Western Blot.Specifications
UBA52 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
UBA52 | |
Recombinant protein fragment corresponding to amino acids 77-128 of human UBA52 (NP_003324) produced in E.coli. | |
0.1 mL | |
7311.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4E1 | |
Western Blot | |
CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction