Learn More
UBA6 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304108A

Description
The recommended shelf life for this product is 1 year from date of receipt.
UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins.UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility.UBA6 is highly expressed in human monocytes.Specifications
UBA6 | |
Polyclonal | |
Unconjugated | |
UBA6 | |
E1-L2, MOP-4, UBE1L2, UBA6, ubiquitin-activating enzyme E1, monocyte protein 4, ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier-activating enzyme 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
231380, 55236 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
A0AVT1, Q8C7R4 | |
UBA6 | |
Between 775 and 825 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.