Learn More
UBA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | UBA6 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8575
![]()
|
Proteintech Group Inc
132111AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes the full-length (118 kDa) and the truncated isoform CRA_b (60-70 kDa) of UBA6 (NCBI).
DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair they function to excise damaged DNA fragments and correct recombinational mismatches. These exonucleases include the family of DNA polymerases. DNA pol alpha, beta, delta and epsilon are involved in DNA replication and repair. DNA pol delta and DNA pol epsilon are multisubunit enzymes, with DNA pol delta consisting of two subunits: p125, which interacts with the sliding DNA clamp protein PCNA; and p50. The nuclear-encoded DNA pol gamma is the only DNA polymerase required for the replication of the mitochondrial DNA. DNA pol zeta is ubiquitously expressed in various tissues and mediates the cellular mechanism of damage-induced mutagenesis. DNA pol theta is a DNA polymerase-helicase that binds ATP and is involved in the repair of interstrand crosslinks.Specifications
| UBA6 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| A0AVT1, Q8C7R4 | |
| 231380, 305268, 55236 | |
| UBA6 Fusion Protein Ag4000 | |
| Primary | |
| -20°C | |
| UBA6 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E1 L2, Monocyte protein 4, MOP 4, MOP4, UBA6, UBE1L2, Ubiquitin activating enzyme 6 | |
| UBA6 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.