Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP181842
Description
UBAC1 Polyclonal specifically detects UBAC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UBAC1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
E3 ubiquitin-protein ligase subunit KPC2, GBDR1UBA domain-containing protein 1, Glialblastoma cell differentiation-related protein 1, Kip1 ubiquitination-promoting complex protein 2, KPC2, putative glialblastoma cell differentiation-related protein, RP11-432J22.3, UBA domain containing 1, UBADC1, ubiquitin associated domain containing 1, ubiquitin-associated domain-containing protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
10422 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
UBAC1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MEMGFDEKEVIDALRVNNNQQNAACEWLLGDRKPSPEELDKGIDPDSPLFQAILDNPVVQLGLTNPKTLLAFEDMLENPLNSTQWMNDPETGPVMLQISRIFQTLNRT | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction