Learn More
UBAC1 Mouse anti-Human, Clone: OTI6D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810287

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.Specifications
UBAC1 | |
Monoclonal | |
Unconjugated | |
UBAC1 | |
GBDR1, KPC2, UBADC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10422 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BSL1 | |
UBAC1 | |
Full length human recombit protein of human UBAC1 produced in 293T. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.